Learn More
Invitrogen™ ZIP13 Polyclonal Antibody

Description
A suggested positive control is K562 cell lysate. PA5-21076 can be used with blocking peptide PEP-1190.
Specifications
Specifications
| Antigen | ZIP13 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SLC39A13 |
| Gene Accession No. | Q96H72 |
| Gene Alias | LIV-1 subfamily of ZIP zinc transporter 9; LZT-Hs9; SLC39A13; solute carrier family 39 (metal ion transporter), member 13; solute carrier family 39 (zinc transporter), member 13; solute carrier family 39 member 13; Zinc transporter ZIP13; ZIP13; ZIP-13; Zrt- and Irt-like protein 13 |
| Gene Symbols | SLC39A13 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.