Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZIP14 Polyclonal Antibody

ZIP14 Polyclonal Antibody
Supplier: Thermo Scientific PA533034
Description
ZIP14 Polyclonal Antibody for IHC (P)
Specifications
| ZIP14 | |
| Polyclonal | |
| Unconjugated | |
| SLC39A14 | |
| cig19; Factor for adipocyte differentiation 123; Fad123; FAD-123; HMNDYT2; Kiaa0062; LIV-1 subfamily of ZIP zinc transporter 4; LZT-Hs4; NET34; SLC39A14; solute carrier family 39 (metal ion transporter), member 14; solute carrier family 39 (zinc transporter), member 14; solute carrier family 39 member 14; Zinc transporter ZIP14; Zip14; ZIP-14; Zrt- and Cig19; Zrt- and Irt-like protein 14; Zrt-, Irt-like protein 14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100152468, 23516 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q15043 | |
| SLC39A14 | |
| Synthetic 15 amino acid peptide from cytoplasmic domain of human SLC39A14. | |
| 50 μg | |
| Primary | |
| Human, Monkey, Pig | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction