Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZIPK Monoclonal Antibody (4H4C8)
GREENER_CHOICE

Catalog No. PIMA531710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531710 Supplier Invitrogen™ Supplier No. MA531710
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31710 has been tested in indirect ELISA.

DAPK3 is a serine/threonine kinase which is functionally linked to cell motility, smooth muscle contractility and cell death processes. DAPK3 is encoded by ZIPK and is a component of desmosomes and of the epidermal cornified envelope in keratinocytes. The N-terminal domain of this protein interacts with the plasma membrane and its C-terminus interacts with intermediate filaments. Through its rod domain, this protein forms complexes with envoplakin. This protein may serve as a link between the cornified envelope and desmosomes as well as intermediate filaments. AKT1/PKB, a protein kinase mediating a variety of cell growth and survival signaling processes, is reported to interact with this protein, suggesting a possible role for this protein as a localization signal in AKT1-mediated signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIPK
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4H4C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene DAPK3
Gene Accession No. O43293
Gene Alias DAP kinase 3; DAP3K; Dapk3; Dapkl; DAP-like kinase; death associated protein kinase 3; death-associated kinase 3; Death-associated like kinase; death-associated protein kinase 3; DLK; MYPT1 kinase; ZIP; ZIP kinase; ZIP kinase isoform; Zipk; ZIP-kinase; zipper-interacting protein kinase
Gene Symbols DAPK3
Host Species Mouse
Immunogen Purified recombinant fragment of human DAPK3 (AA: 28-161) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1613
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.