Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZIPK Polyclonal Antibody
ZIPK Polyclonal Antibody
Supplier: Thermo Scientific PA580539
Description
ZIPK Polyclonal Antibody for ICC/IF, IHC (P)

Specifications
ZIPK | |
Polyclonal | |
Unconjugated | |
DAPK3 | |
DAP kinase 3; DAP3K; Dapk3; Dapkl; DAP-like kinase; death associated protein kinase 3; death-associated kinase 3; Death-associated like kinase; death-associated protein kinase 3; DLK; MYPT1 kinase; ZIP; ZIP kinase; ZIP kinase isoform; Zipk; ZIP-kinase; zipper-interacting protein kinase | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
1613 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with no preservative | |
O43293 | |
DAPK3 | |
A synthetic peptide corresponding to the center region of the Human DAPK3/ZIPK. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction