Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIPK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590193 Supplier Thermo Scientific Supplier No. PA590193
Add to Cart
Add to Cart

ZIPK Polyclonal Antibody

ZIPK Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIPK
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DAPK3
Gene Accession No. O43293, O54784, O88764
Gene Alias DAP kinase 3; DAP3K; Dapk3; Dapkl; DAP-like kinase; death associated protein kinase 3; death-associated kinase 3; Death-associated like kinase; death-associated protein kinase 3; DLK; MYPT1 kinase; ZIP; ZIP kinase; ZIP kinase isoform; Zipk; ZIP-kinase; zipper-interacting protein kinase
Gene Symbols DAPK3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human DAPK3 (NP_0013391).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13144, 1613, 64391
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.