Learn More
ZIPK Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304222AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Regulates both type I apoptotic and type II autophagic cell deaths signal, depending on the cellular setting. The former is caspase- dependent, while the latter is caspase-independent and is characterized by the accumulation of autophagic vesicles. Regulates myosin phosphorylation in both smooth muscle and non- muscle cells. In smooth muscle, regulates myosin either directly by phosphorylating MYL12B and MYL9 or through inhibition of smooth muscle myosin phosphatase (SMPP1M) via phosphorylation of PPP1R12A, and the inhibition of SMPP1M functions to enhance muscle responsiveness to Ca(2+) and promote a contractile state. Enhances transcription from AR-responsive promoters in a hormone- and kinase-dependent manner. Phosphorylates STAT3 and enhances its transcriptional activity. Positively regulates the canonical Wnt/beta-catenin signaling through interaction with NLK and TCF7L2.Specifications
ZIPK | |
Polyclonal | |
Unconjugated | |
DAPK3 | |
DAP3K, ZIP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1613 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O43293 | |
DAPK3 | |
Between 404 and 454 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.