Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CritiCore Protective Wear Zipper Front Reusable Cleanroom Frock

Catalog No. p-7202967
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
6X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Reusable frocks with coil zipper enclosure are suitable for a variety of cleanroom applications including pharmaceutical, biotechnology, and medical device manufacturing.

  • Fluid and bacteria resistant Integrity 1800™ high performance carbon striped fabric
  • Mandarin collar with two snaps at top
  • Wide-pull zipper with a single snap at bottom
  • Cuff Sleeves
  • Laser-cut, pre-serged seams
  • Silicon-free
  • ESD carbon stripes provide electrostatic dissipation

Specifications

Product Type Controlled Environments Frock
Size 2X-Small
Material Polyester
Color White
Disposable Reusable
Sterility Non-sterile
Closure Type Zipper with Snaps
Seam Type Pre-serged
For Use With (Application) Pharmaceutical, Electronics Assembly, Critical Environments
Latex Free Yes
Length (English) 22 in.
Length (Metric) 55.88 cm
Construction Laser-cut, pre-serged seams, silicon-free
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.