Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ziprasidone Hydrochloride Monohydrate 98.0+%, TCI America™
SDP

Catalog No. Z0032250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z0032250MG 250 mg
Z003250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z0032250MG Supplier TCI America Supplier No. Z0032250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 138982-67-9
Molecular Formula C21H24Cl2N4O2S
Molecular Weight (g/mol) 467.409
MDL Number MFCD00921885
InChI Key ZCBZSCBNOOIHFP-UHFFFAOYSA-N
Synonym ziprasidone hydrochloride monohydrate, ziprasidone hydrochloride, geodon, 5-2-4-benzo d isothiazol-3-yl piperazin-1-yl ethyl-6-chloroindolin-2-one hydrochloride hydrate, unii-216x081oru, ziprasidone hcl hydrate, ziprasidone hydrochloride usan:usp, zeldox, ziprasidone hydrochloride usan
PubChem CID 60853
ChEBI CHEBI:32314
IUPAC Name 5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-6-chloro-1,3-dihydroindol-2-one;hydrate;hydrochloride
SMILES C1CN(CCN1CCC2=C(C=C3C(=C2)CC(=O)N3)Cl)C4=NSC5=CC=CC=C54.O.Cl

Specifications

Color White-Yellow
Quantity 250 mg
Formula Weight 449.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Ziprasidone Hydrochloride Monohydrate
RTECSNumber NM3241200
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.