Learn More
Caresfield Ziptop Specimen Bag, 3-Wall with Pouch Tear Away ZipTop Closure

Description
- This size provides ample space for specimen transport, paperwork inserts, and organized sample handling.
- 3-wall construction supports separation and added containment for clinical and laboratory transport workflows.
- Secure ZipTop closure helps reduce leakage risk and supports hygienic specimen handling procedures.
- Clear biohazard bag design allows easy visibility of contents for quick verification during processing.
- Ideal for hospitals, clinics, diagnostic labs, and specimen courier services.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Closure Type | Ziplock |
| Color | Clear |
| Disposable | Single-use |
| Material | Plastic |
| Sterility | Non-sterile |
| Latex Free | Yes |
| Length (English) | 8 in. |
| Width (English) | 10 in. |
| Product Type | Zip Closure Bag |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.