Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zirconium acetate solution

Catalog No. 4138012L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
500 mL
2 L

Catalog No. 41-380-12L

Supplier: Sigma Aldrich 4138012L

Due to product restrictions, please to purchase or view availability for this product.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zirconium acetate solution is a metallic salt solution that can be used for a variety of applications such as: a zirconium based precursor for the development of zirconia coatingsa stabilizer for colloidal solutions, controlled growth of ice crystals, water proofing, and flame retardant treatments

Specifications

CAS 7585-20-8
Density 1.279 g/mL (at 25°C)
Quantity 2 L
Synonym Acetic acid, zirconium salt
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.