Learn More
Zirconium dichloride oxide octahydrate, 98%, Thermo Scientific Chemicals
Description
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.
Solubility
Soluble in water and alcohol.
Notes
Hygroscopic, moisture sensitive and incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 150°C (decomposition) |
Quantity | 500 g |
UN Number | UN3260 |
Merck Index | 14,10184 |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 322.25 (178.13 Anhydrous) |
Percent Purity | 98% |
Odor | Acidic |
Sensitivity | Hygroscopic |
Density | 1.91 |
Show More |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
EINECSNumber | 231-717-9 |
RTECSNumber | ZH7250000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.