Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium fluoride, 99.5%, Thermo Scientific Chemicals

Catalog No. p-221523
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-64-4
Molecular Formula F4Zr
Molecular Weight (g/mol) 167.21
MDL Number MFCD00011307

Specifications

Boiling Point 905.0°C
Melting Point 640.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 25 g
Assay Percent Range 99.4% min
Linear Formula ZrF4
Merck Index 15, 10375
Solubility Information Solubility in water: 1.388 g/100mL. Other solubilities: slightly soluble in hf
Formula Weight 167.21
Percent Purity 99.5%
Packaging Plastic bottle
Chemical Name or Material Zirconium fluoride, 99.5%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
EINECSNumber 232-018-1
RTECSNumber ZH7875000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.