Learn More
Zirconium(IV) bromide, 99%, Thermo Scientific Chemicals
Description
Zirconium(IV) bromide is used as a starting material in the synthesis of zirconium hydride complexes as well as unique cluster compounds. It is also used as a biochemical for proteomics research and as a chemical and organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 450° (15 atm) |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 99% |
Solubility Information | Soluble in water. |
Formula Weight | 410.86 |
Sensitivity | Moisture sensitive |
Density | 4.201 |
Chemical Name or Material | Zirconium(IV) Bromide |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | DOT Class: 8; Packaging Group: III |
EINECSNumber | 237-417-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.