Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium(IV) chloride, 98%, anhydrous, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
10026-11-6
Cl4Zr
233.03
MFCD00011306
Zirconium tetrachloride
10026-11-6
Cl4Zr
233.03
MFCD00011306
Zirconium tetrachloride
Aluminum (Al) 100ppm max.
Assay Percent Range 98%
Linear Formula ZrCl4
Beilstein 01,1295
Fieser 13,16
Merck Index 15,10374
Solubility Information Solubility in water: reacts. Other solubilities: forming hcl and zro2,soluble in alcohol and ether
Formula Weight 233.03
Physical Form Fine Powder
Percent Purity 98%
Chromium (Cr) 100ppm max.
Density 2.8
Hydrogen Fluoride 100ppm max.
Iron (Fe) 1000ppm max.
Packaging Glass bottle
Silicon (Si) 150ppm max.
Solubility Solubility in water: reacts. Other solubilities: forming hcl and zro2, soluble in alcohol and ether
Titanium (Ti) 100ppm max.
Sublimation Point 331 °C
Color White
Melting Point 437.0°C
Quantity 100g
Chemical Name or Material Zirconium(IV) chloride, 98%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
Reacts violently with water.
Corrosive to the respiratory tract.

GHS P Statement
Wear respiratory protection.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
C

GHS Signal Word: Danger

EINECSNumber : 233-058-2

RTECSNumber : ZH7175000

TSCA : TSCA