Learn More
Zirconium(IV) fluoride, 98%, Thermo Scientific Chemicals
Description
Zirconium(IV) fluoride is used as a zirconium source in oxygen-sensitive applications such as metal production. Together with uranium salt, zirconium fluoride can be a component of fuel-coolant in molten salt reactors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 600°C (sublimation) |
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Linear Formula | ZrF4 |
UN Number | UN3260 |
Merck Index | 14,10175 |
Solubility Information | Slightly soluble in water; Soluble in hydrofluoric acid |
Formula Weight | 167.21 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
EINECSNumber | 232-018-1 |
RTECSNumber | ZH7875000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.