Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium(IV) iodide, 99%, Alfa Aesar™

Light, Air and Moisture Sensitive

Manufacturer:  Alfa Aesar 1740014

Quantity 25g

 View more versions of this product

Catalog No. AA1740014


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13986-26-0
99%
598.84
Zirconium tetraiodide
Powder
I4Zr
MFCD00049641

Specifications

13986-26-0
99%
598.84
Zirconium tetraiodide
25g
Powder
I4Zr
MFCD00049641
Yellow
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit