Alfa Aesar™ Zirconium(IV) n-propoxide, 70% w/w in n-propanol

Manufacturer: Alfa Aesar™ 2298918

Quantity 50g

 View more versions of this product

Catalog No. AA2298918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 23519-77-9
    Formula Weight 327.56
    MDL Number MFCD00015307
    UN Number UN1993
    Synonym Tetra-n-propyl zirconate
    Chemical Name or Material Zirconium(IV) n-propoxide

    Specifications

    Formula Weight 327.56
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Solubility Information Hydrolyzes in water
    Physical Form Liquid
    Quantity 50g
    Synonym Tetra-n-propyl zirconate
    Sensitivity Moisture Sensitive
    Health Hazard 1 Danger
    TSCA TSCA
    Density 1.044g/mL
    Concentration 70% w/w in n-propanol
    Refractive Index 1.4510
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    MDL Number MFCD00015307
    CAS 23519-77-9
    UN Number UN1993
    Flash Point 28° (83°F)
    Health Hazard 2 GHS H Statement
    H314-H226-H336
    Causes severe skin burns and eye damage.
    Flammable liquid and vapor.
    May cause drowsiness or dizziness.
    Chemical Name or Material Zirconium(IV) n-propoxide

    GHS H Statement
    H314-H226-H336
    Causes severe skin burns and eye damage.
    Flammable liquid and vapor.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit