Learn More
Zirconium(IV) n-propoxide, 70% w/w in n-propanol, Thermo Scientific Chemicals
Description
Zirconium(IV) n-propoxide is used as a precursor in the production of zirconia. It is also used to study the thermophysical properties like thermal conductivity and thermal effusivity of hybrid films and thereby adjusting optic properties of the films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Liquid |
Quantity | 50 g |
UN Number | UN1993 |
Flash Point | 28°C (83°F) |
Solubility Information | Hydrolyzes in waterImmiscible with water. |
IUPAC Name | propan-1-olate;zirconium(4+) |
Formula Weight | 327.56 |
Concentration or Composition (by Analyte or Components) | 70% w/w in n-propanol |
Refractive Index | 1.451 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H226-H318-H335-H336 |
Hazard Statement | GHS H Statement H314-H226-H336 Causes severe skin burns and eye damage. Flammable liquid and vapor. May cause drowsiness or dizziness. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
EINECSNumber | 245-711-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.