Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zirconium(IV) n-propoxide, 70% w/w in n-propanol, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 4477030

Quantity 250g

 View more versions of this product

Catalog No. AA4477030


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

23519-77-9
327.56
UN1993
Zr[O(CH2)2CH3]4
MFCD00015307
Zirconium (IV) n-propoxide

Specifications

23519-77-9
Liquid
Zr[O(CH2)2CH3]4
MFCD00015307
UN1993
28°C (83°F)
Moisture Sensitive
ChemSeal™ bottles
1.044g/mL
327.56
250g
70% w/w in n-propanol
1.4510
Zirconium (IV) n-propoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H226-H336
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
May cause drowsiness or dizziness.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit