Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium(IV) n-propoxide, 70% w/w in n-propanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog No. p-7059519
Click to view available options
Quantity:
50 g
250 g

Zirconium(IV) n-propoxide is used as a precursor for the preparation of Zirconia. It is used as coupling agent and petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23519-77-9
Molecular Formula C12H28O4Zr
Molecular Weight (g/mol) 327.58
MDL Number MFCD00015307
InChI Key XPGAWFIWCWKDDL-UHFFFAOYSA-N
Synonym zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
PubChem CID 90139
IUPAC Name propan-1-olate;zirconium(4+)
SMILES CCCO[Zr](OCCC)(OCCC)OCCC

Specifications

Density 1.044 g/mL
Flash Point 28°C (83°F)
Packaging ChemSeal™ bottles
Refractive Index 1.451
Quantity 50 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Not miscible in water.
Formula Weight 327.56
Concentration or Composition (by Analyte or Components) 70% w/w in n-propanol
Physical Form Liquid
Chemical Name or Material Zirconium (IV) n-propoxide
Show More Show Less
Hazard Category H226-H318-H335-H336
Hazard Statement GHS H Statement
H314-H226-H336
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 245-711-9
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.