Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium(IV) oxide 2-ethylhexanoate, in mineral spirits (≈6% Zr), Thermo Scientific Chemicals

Catalog No. p-7050816
Click to view available options
Quantity:
500 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22464-99-9
Molecular Formula C8H16O2Zr
Molecular Weight (g/mol) 235.438
MDL Number MFCD00072684
InChI Key HBHXRPHNNKAWQL-UHFFFAOYSA-N
Synonym zirconium 2-ethylhexanoate
PubChem CID 73555628
IUPAC Name 2-ethylhexanoic acid;zirconium
SMILES CCCCC(CC)C(=O)O.[Zr]

Specifications

Flash Point 40°C (104°F)
Quantity 500 g
UN Number UN1268
Concentration or Composition (by Analyte or Components) ≈6% Zr
Physical Form Liquid
Chemical Name or Material Zirconium(IV) oxide 2-ethylhexanoate, In mineral spirits
Hazard Category H226-H302-H336-H372
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P314-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.
TSCA Yes

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.