Learn More
Zirconium n-butoxide, 80% w/w in 1-butanol, Thermo Scientific Chemicals
Description
Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Zirconium(IV) butoxide solution used in the preparation of CeO2-stabilized zirconia polycrystals gel that have good applications as protective coatings of steel and ferrous alloys.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from oxidizing agents, acids, bases, halogens, alkali metals, water/ moisture.
Specifications
Specifications
| Quantity | 250 g |
| Linear Formula | [CH3(CH2)3O]4Zr |
| UN Number | UN1120 |
| Beilstein | 4132649 |
| Flash Point | 38°C (100°F) |
| Solubility Information | Hydrolyzes in water. |
| Formula Weight | 383.68 |
| Concentration or Composition (by Analyte or Components) | 80% w/w in 1-butanol |
| Odor | Alcohol-like |
| Refractive Index | 1.466 |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H318-H335-H336 |
| Hazard Statement | GHS H Statement H318-H226-H335-H336-H315 Causes serious eye damage. Flammable liquid and vapour. May cause respiratory irritation. May cause drowsiness or dizziness. Causes skin irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS |
| EINECSNumber | 213-995-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.