Learn More
Zirconium titanium oxide, 99.5% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals
Description
Suitable for glass, optic and ceramic applications, advanced electronics and in light weight structural components in aerospace and electrochemical applications such as fuel cells in which they exhibit ionic conductivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 100 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 203.11 |
| Percent Purity | 99.5% |
| Odor | Odorless |
| Assay | (metals basis excluding Hf), Hf <75ppm |
| Chemical Name or Material | Zirconium titanium oxide |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.