Learn More
COAST Cutlery Zithion-X USB-C Ported Battery
Pack power with purpose. COAST ZITHION-X™ rechargeable lithium batteries change the game in responsible, portable power technology.
$29.99
Specifications
Product Type | Replacement Battery |
---|---|
Material | Lithium-ion |
Quantity | 4/Pk. |
Voltage | 1.5 V |
Catalog Number | Mfr. No. | Battery Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Battery Type | Price | Quantity & Availability | |||||
19-030-059
|
COAST Cutlery
31007 |
AA |
Each for $29.99
|
|
|||||
19-030-060
|
COAST Cutlery
31008 |
AAA |
Each for $29.99
|
|
|||||
Description
- Battery Charging Level Indicator - Check the status of your batteries charge level with our built-in charge level indicator. (Solid red) when charging, (solid green) when fully charged.
- Built-In USB-C Port - With a built-in USB-C port, your battery can be charged using a USB-C cord. Some COAST models can be charged both inside or outside the light.
- Overcharge/Discharge Protection Circuits - Over charging and improperly discharging lithium batteries can cause a safety concern and hazardous risk to the user and it's surroundings. COAST batteries are built with overcharge/discharge protection, ensuring you and your battery are protected.
Specifications
Replacement Battery | |
Lithium-ion | |
4/Pk. | |
1.5 V |