Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZKSCAN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ZKSCAN1 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ZKSCAN1 Polyclonal specifically detects ZKSCAN1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ZKSCAN1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
7586 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:NLARRNLSRDNRQENYGSAFPQGGENRNENEESTSKAETSEDSASRGETTGRSQKEFGEKRDQEGKTGERQQKNPEEKTRKEKRDSGPAIGKDKKTITGERGPREKGKGLGRSFSLSSNFTTPEEVPTGTKSHRCDECGKCFTR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
Polyclonal | |
Rabbit | |
Human | |
KOX189130423L19Rik, PHZ-37, Zinc finger protein 139, Zinc finger protein 36, zinc finger protein 36 (KOX 18), Zinc finger protein KOX18, zinc finger protein with KRAB and SCAN domains 1, zinc finger with KRAB and SCAN domains 1, ZNF139, ZNF36MGC138429 | |
ZKSCAN1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title