Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZKSCAN1 Monoclonal Antibody (2C7)
GREENER_CHOICE

Catalog No. PIMA520255
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520255 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520255 Supplier Invitrogen™ Supplier No. MA520255
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MMTAESREAT GLSPQAAQEK DGIVIAKVEE EDEEDHMWGQ DSTLQDTPPP DPEIFRQRFR RFCYQNTFGP REALSRLKEL CHQWLRPEIN TKEQILELLV LEQFLSILPK ELQVWLQEYR PDSGEEAVTL LEDLELDLSG QQVPGQVHGP EMLARGMVPL DPVQESSSFD LHHEATQSHF KHSSRKPRLL QSRALPAAHI PAPPHEGSPR DQAMASALFT ADSQAMVKIE DMAVSLILEE WGCQNLARRN LSRDNRQENY GSAFPQGGEN RNENEESTSK AETSEDSASR GETTGRSQKE FGEKRDQEGK TGERQQKNPE EKTRKEKRDS GPAIGKDKKT ITGERGPREK GKGLGRSFSL SSNFTTPEEV PTGTKSHRCD ECGKCFTRSS SLIRHKIIHT GEKPYECSEC GKAFSLNSNL VLHQRIHTGE KPHECNECGK AFSHSSNLIL HQRIHSGEKP YECNECGKAF SQSSDLTKHQ RIHTGKAFNR NSYLILHRRI HTREKPYKCT KCGKAFTRSS TLTLHHRIHA RERASEYSPA SLDAFGAFLK SCV.

The ZKSCAN1 gene encodes a transcriptional regulator of the KRAB. Transcriptional regulatory proteins containing tandemly repeated zinc finger domains are thought to be involved in both normal and abnormal cellular proliferation and differentiation. See ZNF91 (MIM 603971) for general information on zinc finger proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZKSCAN1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C7
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZKSCAN1
Gene Accession No. P17029
Gene Alias 5930429A01Rik; 9130423L19Rik; 9230118B16Rik; AI646829; AI788588; C87323; KOX18; LOC100720521; PHZ-37; zinc finger and SCAN domain containing 21; zinc finger and SCAN domain-containing protein 21; zinc finger protein 139; Zinc finger protein 36; zinc finger protein 36 (KOX 18); zinc finger protein KOX18; Zinc finger protein with KRAB and SCAN domains 1; zinc finger with KRAB and SCAN domains 1; Zkscan1; ZNF139; ZNF36; Zscan21; ZSCAN33
Gene Symbols ZKSCAN1
Host Species Mouse
Immunogen ZKSCAN1 (AAH22378, 1 a.a. ~ 563 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7586
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.