Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZKSCAN1 Monoclonal Antibody (OTI3C4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF810967


Description
ZKSCAN1 Monoclonal antibody specifically detects ZKSCAN1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
ZKSCAN1 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q64305, Q7RTS3, Q9QX98 | |
Mouse | |
Protein A/G | |
RUO | |
117034, 19213, 256297 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3C4 | |
Unconjugated | |
PTF1A | |
5930429A01Rik; 9130423L19Rik; 9230118B16Rik; AI646829; AI788588; C87323; KOX18; LOC100720521; PHZ-37; zinc finger and SCAN domain containing 21; zinc finger and SCAN domain-containing protein 21; zinc finger protein 139; Zinc finger protein 36; zinc finger protein 36 (KOX 18); zinc finger protein KOX18; Zinc finger protein with KRAB and SCAN domains 1; zinc finger with KRAB and SCAN domains 1; Zkscan1; ZNF139; ZNF36; Zscan21; ZSCAN33 | |
Full length human recombinant protein of human PTF1A produced in E.coli. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction