Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZKSCAN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552321 Supplier Invitrogen™ Supplier No. PA552321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLARRNLSRD NRQENYGSAF PQGGENRNEN EESTSKAETS EDSASRGETT GRSQKEFGEK RDQEGKTGER QQKNPEEKTR KEKRDSGPAI GKDKKTITGE RGPREKGKGL GRSFSLSSNF TTPEEVPTGT KSHRCDECGK CFTR Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.

The ZKSCAN1 gene encodes a transcriptional regulator of the KRAB. Transcriptional regulatory proteins containing tandemly repeated zinc finger domains are thought to be involved in both normal and abnormal cellular proliferation and differentiation. See ZNF91 (MIM 603971) for general information on zinc finger proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZKSCAN1
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZKSCAN1
Gene Accession No. P17029
Gene Alias 5930429A01Rik; 9130423L19Rik; 9230118B16Rik; AI646829; AI788588; C87323; KOX18; LOC100720521; PHZ-37; zinc finger and SCAN domain containing 21; zinc finger and SCAN domain-containing protein 21; zinc finger protein 139; Zinc finger protein 36; zinc finger protein 36 (KOX 18); zinc finger protein KOX18; Zinc finger protein with KRAB and SCAN domains 1; zinc finger with KRAB and SCAN domains 1; Zkscan1; ZNF139; ZNF36; Zscan21; ZSCAN33
Gene Symbols ZKSCAN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZKSCAN1. Recombinant protein control fragment (Product #RP-89318).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7586
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.