Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZKSCAN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ZKSCAN1 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4790
![]() ![]() |
Proteintech Group Inc
252141AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The ZKSCAN1 gene encodes a transcriptional regulator of the KRAB. Transcriptional regulatory proteins containing tandemly repeated zinc finger domains are thought to be involved in both normal and abnormal cellular proliferation and differentiation. See ZNF91 (MIM 603971) for general information on zinc finger proteins.Specifications
ZKSCAN1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P17029, Q8BGS3 | |
74570, 7586 | |
ZKSCAN1 Fusion Protein Ag19721 | |
Primary | |
-20°C | |
ZKSCAN1 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
9130423L19Rik, KOX18, PHZ 37, Zinc finger protein 139, Zinc finger protein 36, Zinc finger protein KOX18, ZKSCAN1, ZNF139, ZNF36 | |
Zkscan1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title