Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZKSCAN2 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305894C
Description
ZKSCAN2 Polyclonal specifically detects ZKSCAN2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ZKSCAN2 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
zinc finger protein with KRAB and SCAN domains 2, zinc finger with KRAB and SCAN domains 2, ZNF694, ZSCAN31 | |
Produced in rabbits immunized with E. coli-derived Human ZKSCAN2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
342357 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction