Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZKSCAN3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP247570
Description
ZKSCAN3 Polyclonal specifically detects ZKSCAN3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZKSCAN3 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
dJ874C20.1, FLJ33906, ZF47, Zfp47, Zfp-47, Zinc finger and SCAN domain-containing protein 13, Zinc finger protein 306KIAA0426, Zinc finger protein 309ZNF306ZFP306, Zinc finger protein 47 homolog, zinc finger protein with KRAB and SCAN domains 3, zinc finger protein zfp47, zinc finger with KRAB and SCAN domains 3, ZNF309, ZSCAN13 | |
Rabbit | |
Affinity Purified | |
RUO | |
80317 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ZKSCAN3 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LLTPAPGSQSSQFQLMKALLKHESVGSQPLQDRVLQVPVLAHGGCCREDKVVASRLTPESQGLLKVEDVALTLTPEWTQQDSSQGNLCRDEKQENHGSLVSLGDEKQTKSRDLPPAEELPEKEHGKISCHLREDIAQIPTCAEAGEQEGR | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction