Learn More
Invitrogen™ ZKSCAN4 Monoclonal Antibody (1D2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA523296
Description
Peptide Sequence: MAREPRKNAA LDAQSAEDQT GLLTVKVEKE EASALTAEVR APCSPARGPE RSRQRFRGFR YPEAAGPREA LSRLRELCGQ WLQPEMHSKE QILELLVLEQ FLTILPGNLQ SWVREQHPES GEEVVVLLEY LERQLDEPAP QVPVGDQGQE LLCCKMALLT QTQGSQSSQC QPMKALFKHE SLGSQPLHDR VLQVPGLAQG GCCREDAMVA SRLTPGSQGL LKMEDVALTL TPGWTQLDSS QVNLYRDEKQ ENHSSLVSLG GEIQTKSRDL PPVKKLPEKE HGKICHLRED IAQIPTHAEA GEQEGRLQRK QKNAIGSRRH YCHECGKSFA QSSGLTKHRR IHTGEKPYEC EDCGKTFIGS SALVIHQRVH TGEKPYECEE CGKVFSHSSN LIKHQRTHTG EKPYECDDCG KTFSQSCSLL EHHKIHTGEK PYQCNMCGKA FRRNSHLLRH QRIHGDKNVQ NPEHGESWES QGRTESQWEN TEAPVSYKCN ECERSFTRNR SLIEHQKIHT GEKPYQCDTC GKGFTRTSYL VQHQRSHVGK KTLSQ.
ZKSCAN4 may be involved in the transcriptional activation of MDM2 and EP300 genes.
Specifications
| ZKSCAN4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q969J2 | |
| ZKSCAN4 | |
| ZKSCAN4 (AAH14031, 1 a.a. ~ 545 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 1D2 | |
| Unconjugated | |
| ZKSCAN4 | |
| AI841875; Gm11272; OTTMUSG00000000421; P1P373C6; P373c6.1; p373c6.1 (novel C2H2 type zinc finger protein); Zfp307; Zinc finger protein 307; zinc finger protein 427; Zinc finger protein with KRAB and SCAN domains 4; zinc finger with KRAB and SCAN domains 4; ZKSCAN4; ZNF307; ZNF427; ZSCAN36 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 387032 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.