Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZM-306416 hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number AAJ67265MCR
Click to view available options
Quantity:
1 mg
10 mg
690206-97-4
C16H13ClFN3O2
333.747
MFCD08703133
YHUIUSRCUKUUQA-UHFFFAOYSA-N
n-4-chloro-2-fluorophenyl-6,7-dimethoxyquinazolin-4-amine, anilinoquinazoline deriv. 4, d0c6ki, n-2-fluoro-4-chlorophenyl-6,7-dimethoxyquinazoline-4-amine, n-4-chloro-2-fluorophenyl-6,7-dimethoxy-4-quinazolinamine
5329006
N-(4-chloro-2-fluorophenyl)-6,7-dimethoxyquinazolin-4-amine
COC1=C(C=C2C(=C1)C(=NC=N2)NC3=C(C=C(C=C3)Cl)F)OC
690206-97-4
C16H13ClFN3O2
333.747
MFCD08703133
YHUIUSRCUKUUQA-UHFFFAOYSA-N
n-4-chloro-2-fluorophenyl-6,7-dimethoxyquinazolin-4-amine, anilinoquinazoline deriv. 4, d0c6ki, n-2-fluoro-4-chlorophenyl-6,7-dimethoxyquinazoline-4-amine, n-4-chloro-2-fluorophenyl-6,7-dimethoxy-4-quinazolinamine
5329006
N-(4-chloro-2-fluorophenyl)-6,7-dimethoxyquinazolin-4-amine
COC1=C(C=C2C(=C1)C(=NC=N2)NC3=C(C=C(C=C3)Cl)F)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 1 mg
Formula Weight 370.21
Percent Purity 98%
Physical Form Solid
Chemical Name or Material ZM-306416 hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit