Learn More
ZM 39923 hydrochloride, Tocris Bioscience™
Description
Specifications
Specifications
| Quantity | 10 mg |
| Synonym | zm 39923 hydrochloride, jak3 inhibitor iv, zm39923 hydrochloride, 3-benzyl isopropyl amino-1-naphthalen-2-yl propan-1-one hydrochloride, 3-benzylisopropylamino-1-naphthalen-2-yl-propan-1-one hydrochloride, 2-naphthyl-n-isopropyl,n-benzyl-beta-aminoethylketone, hci, 3-n-benzyl-n-isopropyl amino-1-naphthalen-2-yl propan-1-one hydrochloride, c23h25no.clh |
| Formula Weight | 367.92 |
| Percent Purity | >99% |
| Chemical Name or Material | ZM 39923 hydrochloride |
Safety and Handling
| Recommended Storage | Desiccate at +4°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.