Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ZM 447439
GREENER_CHOICESDP

Catalog No. 245810 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-581-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 24-581-0 Supplier Tocris Bioscience™ Supplier No. 2458/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibits Aurora kinase B

Novel, selective ATP-competitive inhibitor of Aurora B kinase in vitro (IC50 values are 50, 250 and 1000 nM for Aurora B, C and A kinases respectively). Selective over a range of other kinases including cdk1 and PLK1 (IC50 > 10 μM). Inhibits cell division and displays selective toxicity towards proliferating tumor cells versus non-dividing cells.

Chemical Identifiers

CAS 331771-20-1
Molecular Formula C29H31N5O4

Specifications

Quantity 10 mg
Formula Weight 513.59
Purity >99%
Molecular Weight (g/mol) 513.598
Chemical Name or Material ZM 447439
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.