Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552991
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552991 Supplier Invitrogen™ Supplier No. PA552991
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSRKTQDSYQ NECADYINVQ KARGLEAKTC FRKMEESSLE TRRYREVVDS RPRHRMFEQR LPFETFRTYA APYNISQAME KQLPHSKKTY DSFQDELEDY IKVQKARGLD PKTCFRKMRE NSVDTHGYRE MVDSGP Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 55%.

This gene encodes a protein containing Cys2-His2 (C2H2)-type zinc fingers, which are similar to those found in the nuclear matrix protein matrin 3. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Jan 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMAT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZMAT1
Gene Accession No. Q5H9K5
Gene Alias A230096A06; AW990744; B930008K04Rik; KIAA1789; RGD1565344; zinc finger matrin-type 1; zinc finger matrin-type 1; zinc finger matrin-type protein 1; zinc finger matrin-type protein 1; zinc finger, matrin type 1; zinc finger, matrin-type 1; ZMAT1
Gene Symbols ZMAT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZMAT1. Recombinant protein control fragment (Product #RP-92230).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84460
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.