Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567539
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567539 Supplier Invitrogen™ Supplier No. PA567539
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specificity: The antibody detects endogenous levels of total ZMAT2 protein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMAT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Zmat2
Gene Accession No. Q96NC0, Q9CPW7
Gene Alias 2610510D14Rik; 2900082I05Rik; Ptg-12; Snu23; zinc finger matrin-type 2; zinc finger matrin-type protein 2; zinc finger, matrin type 2; zinc finger, matrin-type 2; ZMAT2
Gene Symbols Zmat2
Host Species Rabbit
Immunogen Full length Fusion protein of human ZMAT2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153527, 66492
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.