Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589776
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589776 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589776 Supplier Invitrogen™ Supplier No. PA589776
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASGSGTKNL DFRRKWDKDE YEKLAEKRLT EEREKKDGKP VQPVKRELLR HRDYKVDLES KLGKTIVITK TTPQSEMGGY YCNVCDCVVK DSINFLDHIN GKKHQRNLGM SMRVERSTLD QVKKRFEVNK KKMEEKQKDY DFEERMKELR EEEEKAKAYK KEKQKEKKRR AEEDLTFEED DEMAAVMGFS GFGSTKKSY; Positive Samples: LO2, Jurkat, HeLa, SH-SY5Y, MCF7, Mouse brain, Rat brain; Cellular Location: Nucleus.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMAT2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Zmat2
Gene Accession No. Q96NC0, Q9CPW7
Gene Alias 2610510D14Rik; 2900082I05Rik; Ptg-12; Snu23; zinc finger matrin-type 2; zinc finger matrin-type protein 2; zinc finger, matrin type 2; zinc finger, matrin-type 2; ZMAT2
Gene Symbols Zmat2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-199 of human ZMAT2 (NP_6533241).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153527, 66492, 679898
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.