Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMAT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582985 Supplier Invitrogen™ Supplier No. PA582985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GPYFNPRSRQ RIPRDLAMCV TPSGQFYCSM CNVGAGEEME FRQHLESKQH KSKVSEQRYR NEMENLGYV Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.

This gene encodes a protein containing three zinc finger domains and a nuclear localization signal. The mRNA and the protein of this gene are upregulated by wildtype p53 and overexpression of this gene inhibits tumor cell growth, suggesting that this gene may have a role in the p53-dependent growth regulatory pathway. Alternative splicing of this gene results in two transcript variants encoding two isoforms differing in only one amino acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMAT3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZMAT3
Gene Accession No. Q9HA38
Gene Alias FLJ12296; matrin type 3; MGC10613; p53 target zinc finger protein; p53-activated gene 608 protein; Pag608; UniProtKB:Q9HA38}; Wig1; WIG-1; WIG-1/PAG608 protein; wild-type p53-induced gene 1; wild-type p53-induced gene 1 protein; zinc finger matrin type 3; zinc finger matrin-type 3; zinc finger matrin-type protein 3; zinc finger protein WIG1; Zinc finger protein WIG-1; zinc finger, matrin type 3; zmat3; zmat3 {ECO:0000250
Gene Symbols ZMAT3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZMAT3. Recombinant protein control fragment (Product #RP-106535).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64393
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.