Learn More
Invitrogen™ ZMAT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596307
Description
Immunogen sequence: MILLQHAVLP PPKQPSPSPP MSVATRSTGT LQLPPQKPFG QEASLPLAGE EELSKGGEQD CALEELCKPL YCKLCNVTLN SAQQAQAHYQ GKNHGKKLRN YYAANSCPPP ARMSNVVEPA ATPVVPVPPQ MGSFKPGGRV ILATENDYCK LCDASFSSPA VAQAHYQGKN HAKRLRLAEA QSNSFSESSE LGQRRARKEG NEFKMMPNRR NMYTVQNNSA GPYFNPRSRQ RIPRDLAMCV TPSGQFYCSM CNVGAGEEME FRQHLESKQH KSKVSEQRYR NEMENLGYV; Positive Samples: Mouse brain, Rat brain; Cellular Location: Nucleus, nucleolus.
This gene encodes a protein containing three zinc finger domains and a nuclear localization signal. The mRNA and the protein of this gene are upregulated by wildtype p53 and overexpression of this gene inhibits tumor cell growth, suggesting that this gene may have a role in the p53-dependent growth regulatory pathway. Alternative splicing of this gene results in two transcript variants encoding two isoforms differing in only one amino acid.
Specifications
ZMAT3 | |
Polyclonal | |
Unconjugated | |
ZMAT3 | |
FLJ12296; matrin type 3; MGC10613; p53 target zinc finger protein; p53-activated gene 608 protein; Pag608; UniProtKB:Q9HA38}; Wig1; WIG-1; WIG-1/PAG608 protein; wild-type p53-induced gene 1; wild-type p53-induced gene 1 protein; zinc finger matrin type 3; zinc finger matrin-type 3; zinc finger matrin-type protein 3; zinc finger protein WIG1; Zinc finger protein WIG-1; zinc finger, matrin type 3; zmat3; zmat3 {ECO:0000250 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22401, 64393, 64394 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.62 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08781, O54836, Q9HA38 | |
ZMAT3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-289 of human ZMAT3 (NP_0719151). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.