Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMAT4 Monoclonal Antibody (OTI9E11)
GREENER_CHOICE

Catalog No. PIMA527196
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527196 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527196 Supplier Invitrogen™ Supplier No. MA527196
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZMAT4 Monoclonal Antibody for Western Blot

ZMAT4 (Zinc Finger Matrin-Type 4) is a Protein Coding gene. Diseases associated with ZMAT4 includeRefractive Error. Gene Ontology (GO) annotations related to this gene includenucleic acid binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMAT4
Applications Western Blot
Classification Monoclonal
Clone OTI9E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZMAT4
Gene Accession No. Q9H898
Gene Alias 9630048M01Rik; SDP1; zinc finger matrin-type 4; zinc finger matrin-type protein 4; zinc finger, matrin type 4; zinc finger, matrin-type 4; Zmat4
Gene Symbols ZMAT4
Host Species Mouse
Immunogen Full length human recombinant protein of ZMAT4 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79698
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.