Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZMAT4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564430
Description
Immunogen sequence: QHYDGKKHKK NAARVALLEQ LGTTLDMGEL RGLRRNYRCT ICSVSLNSIE QYH Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 84%.
ZMAT4 (Zinc Finger Matrin-Type 4) is a Protein Coding gene. Diseases associated with ZMAT4 includeRefractive Error. Gene Ontology (GO) annotations related to this gene includenucleic acid binding.
Specifications
ZMAT4 | |
Polyclonal | |
Unconjugated | |
ZMAT4 | |
9630048M01Rik; SDP1; zinc finger matrin-type 4; zinc finger matrin-type protein 4; zinc finger, matrin type 4; zinc finger, matrin-type 4; Zmat4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79698 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H898 | |
ZMAT4 | |
Recombinant protein corresponding to Human ZMAT4. Recombinant protein control fragment (Product #RP-104074). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction