Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMAT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552123
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552123 Supplier Invitrogen™ Supplier No. PA552123
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKKHLNGLQH LKAKKVWYDM FRDAAAILLD EQNKRPCRKF LLTGQCDFGS NCRFSHMSER DLQELSIQVE EERRAREWLL DAPELPEGHL EDWLEKRAKR LSSAPS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMAT5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZMAT5
Gene Accession No. Q9UDW3
Gene Alias SNRNP20; U11/U12 small nuclear ribonucleoprotein 20 kDa protein; U11/U12 snRNP 20 kDa protein; U11/U12 snRNP 20K; U11/U12-20K; ZC3H19; zinc finger CCCH-type containing 19; zinc finger matrin-type 5; zinc finger matrin-type protein 5; zinc finger, matrin-type 5; ZMAT5
Gene Symbols ZMAT5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZMAT5. Recombinant protein control fragment (Product #RP-91543).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55954
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.