Learn More
Invitrogen™ ZMIZ1 Polyclonal Antibody

Description
Immunogen sequence: MNSMDRHIQQ TNDRLQCIKQ HLQNPANFHN AATELLDWCG DPRAFQRPFE QSLMGCLTVV SRVAAQQGFD LDLGYRLLAV CAANRDKFT Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Specifications
Specifications
| Antigen | ZMIZ1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZMIZ1 |
| Gene Accession No. | Q9ULJ6 |
| Gene Alias | BC065120; E330020C23; ENSMUSG00000072684; FLJ00092 protein; Gm10397; hZIMP10; I920194n01; KIAA1224; MIZ; PIAS-like protein Zimp10; Rai17; retinoic acid induced 17; retinoic acid-induced protein 17; RP11-519K18.1; TRAFIP10; Zimp10; Zinc finger MIZ domain-containing protein 1; zinc finger MIZ-type containing 1; zinc finger, MIZ-type containing 1; zinc finger-containing, Miz1, PIAS-like protein on chromosome 10; ZMIZ1 |
| Gene Symbols | ZMIZ1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.