Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMIZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140136 Supplier Invitrogen™ Supplier No. PA5140136
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: EPNYGNQQYG PNSQFPTQPG QYPAPNPPRP LTSPNYPGQR MPSQPSSGQY PPPTVNMGQY YKPEQFNGQN NTFSGSSYSN YSQGNV Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 94%.

Retinoic acid plays a critical role in development, cellular growth, and differentiation and induces the expression of a variety of genes. RAI17 expression is induced by retinoic acid and is predominantly expressed in heart, brain and ovaries. Within brain, highest expression is in amygdala. The deduced 1,067-amino acid protein contains an MSX-interacting zinc finger (MIZ) domain, a nuclear localization signal sequence, and 2 proline-rich regions. A strong intrinsic transactivation domain is identified in the C-terminal proline-rich region. RAI17 expression is detected in various cancer cell lines. RAI17 colocalizes with endogenous androgen receptor (AR) in the nuclei of prostate epithelial cells from human tissue samples. In human prostate cancer cells, RAI17 increases the transcriptional activity of AR. Studies using sumoylation-deficient AR mutants suggest that the increase of AR activity by RAI17 is dependent upon receptor sumoylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMIZ1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZMIZ1
Gene Accession No. Q9ULJ6
Gene Alias BC065120; E330020C23; ENSMUSG00000072684; FLJ00092 protein; Gm10397; hZIMP10; I920194n01; KIAA1224; MIZ; PIAS-like protein Zimp10; Rai17; retinoic acid induced 17; retinoic acid-induced protein 17; RP11-519K18.1; TRAFIP10; Zimp10; Zinc finger MIZ domain-containing protein 1; zinc finger MIZ-type containing 1; zinc finger, MIZ-type containing 1; zinc finger-containing, Miz1, PIAS-like protein on chromosome 10; ZMIZ1
Gene Symbols ZMIZ1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZMIZ1. Recombinant protein control fragment (Product #RP-109251).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57178
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.