Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZMIZ1/Zimp10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19261025UL
Description
ZMIZ1/Zimp10 Polyclonal specifically detects ZMIZ1/Zimp10 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ZMIZ1/Zimp10 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
FLJ13541, hZIMP10, KIAA1224Zimp10, MIZ, PIAS-like protein Zimp10, RAI17TRAFIP10, retinoic acid induced 17, Retinoic acid-induced protein 17, RP11-519K18.1, ZIMP10FLJ00092 protein, zinc finger MIZ domain-containing protein 1, zinc finger, MIZ-type containing 1, zinc finger-containing, Miz1, PIAS-like protein on chromosome 10 | |
Rabbit | |
Affinity Purified | |
RUO | |
57178 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ZMIZ1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MNSMDRHIQQTNDRLQCIKQHLQNPANFHNAATELLDWCGDPRAFQRPFEQSLMGCLTVVSRVAAQQGFDLDLGYRLLAVCAANRDKFT | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction