Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZMIZ2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145018
Description
Immunogen sequence: SEVYPGQQYL QGGQYAPSTA QFAPSPGQPP APSPSYPGHR LPLQQGMTQS LSVPGPTGLH YKPTEQFNGQ Antigen sequence identity: Mouse 86%; Rat 87%.
ZIMP7, also known as ZMIZ2, is a novel PIAS (protein inhibitor of activated signal transducer and activator of transcription)-like protein and a transcriptional coactivator. ZIMP7 is expressed most abundantly in testis. The C-terminal proline-rich domain possesses a significant intrinsic transcriptional activity and this activity is inhibited by the N-terminus in the full-length ZIMP7. ZIMP7 and the related protein ZIMP10 interact with PIAS3 and enhances Androgen Receptor (AR)- mediated transcription. The interaction between ZIMP7 and SWI/SNF complex suggests a possible role for ZIMP7 in chromatin modification.
Specifications
ZMIZ2 | |
Polyclonal | |
Unconjugated | |
ZMIZ2 | |
D11Bwg0280e; HRIHFB2007; hZIMP7; KIAA1886; NET27; PIAS-like protein Zimp7; TRAFIP20; Zimp7; zinc finger MIZ domain-containing protein 2; zinc finger MIZ-type containing 2; zinc finger, MIZ-type containing 2; Zmiz2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
83637 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NF64 | |
ZMIZ2 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 270-339. Recombinant protein control fragment (Product #RP-109798). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction