Learn More
Invitrogen™ ZMPSTE24 Polyclonal Antibody

Description
ZMPSTE24 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | ZMPSTE24 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ZMPSTE24 |
| Gene Accession No. | O75844 |
| Gene Alias | A530043O15Rik; CAAX prenyl protease 1 homolog; D030046F19; Face1; Face-1; Farnesylated proteins-converting enzyme 1; farnesylated-proteins converting enzyme 1; fi45a02; FLJ14968; HGPS; MADB; Prenyl protein-specific endoprotease 1; PRO1; STE24; Ste24p; wu:fi45a02; zgc:55655; zinc metallopeptidase STE24; zinc metallopeptidase STE24 homolog; zinc metallopeptidase, STE24; zinc metallopeptidase, STE24 homolog; zinc metalloproteinase (STE24 homolog, yeast); zinc metalloproteinase Ste24 homolog; zinc metalloproteinase, STE24 homolog; Zmpste24 |
| Gene Symbols | ZMPSTE24 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.