Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMYM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566658
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566658 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566658 Supplier Invitrogen™ Supplier No. PA566658
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSTQIQSDIIE IIKTEMLQDI VNEINDSSAF SIICDETINS AMKEQLSICV RYPQKSSKAI LIKERFLGFV DTEEMTGTHL Highest antigen sequence identity to the following orthologs - mouse 67%, rat 64%.

Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. As a member of the MYM (myeloproliferative and mental retardation) gene family, ZMYM1 is widely expressed in different tissues in eukaryotes under several forms derived by alternative splicing. While its function remains unknown, the related protein ZMYM2 has been shown to associate with and stabilize the LSD1-CoREST-HDAC1 (LCH) complex of chromatin through its MYM-type zinc fingers, thereby enhancing the transcriptional repression of several genes, suggesting that ZMYM1 may play a similar role.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMYM1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZMYM1
Gene Accession No. Q5SVZ6
Gene Alias 5830412B09Rik; AW209050; MYM; RP11-181E22.4; RP11-248I9.4; zinc finger MYM-type containing 1; zinc finger MYM-type protein 1; zinc finger, MYM domain containing 1; zinc finger, MYM-type 1; ZMYM1
Gene Symbols ZMYM1
Host Species Rabbit
Immunogen Recombinant Human ZMYM1. Recombinant protein control fragment (Product #RP-107306).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79830
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.