Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMYM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583208 Supplier Invitrogen™ Supplier No. PA583208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLSFGTVFRH WKKNPLTMEN KACLRYQVSS LCGTDNEDKI TTGKRKHEDD EPVFEQIENT ANPSRCPVKM FECYLSKSPQ N Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.

ZNF198 is a ubiquitously expressed nuclear protein whose physiological role is largely unknown. It consists of 5 N-terminal zinc finger motifs responsible for protein-protein interactions, a central proline-rich domain and C-terminal nuclear localization signal (NLS) and an acidic domain. Reciprocal chromosomal translocation involving ZNF198 and FGFR1 results in the formation of ZNF198/FGFR1 fusion kinase protein. This fusion protein is a ligand-dependent, constitutively active cytoplasmic tyrosine kinase and regulates several STAT transcription factors including STAT 1, 3 and 5. This fusion protein is an oncogenic protein and is associated with an atypical myeloproliferative disease, peripheral blood eosinophilia and T-cell leukemia/lymphoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMYM2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZMYM2
Gene Accession No. Q9UBW7
Gene Alias 5830413P05Rik; FIM; Fused in myeloproliferative disorders protein; MYM; RAMP; rearranged in an atypical myeloproliferative disorder; Rearranged in atypical myeloproliferative disorder protein; RP11-264J4.7; SCLL; Zfp198; zinc finger MYM-type containing 2; zinc finger MYM-type protein 2; Zinc finger protein 198; zinc finger, MYM-type 2; Zmym2; ZNF198
Gene Symbols ZMYM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZMYM2. Recombinant protein control fragment (Product #RP-95485).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7750
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.